Heat sealing foil, ThermASeal Foil™
Leverandør: iST Scientific Ltd
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
IST-108-115TR IST-108-078LR IST-108-078LS IST-108-078SR IST-108-078SS IST-108-078TR IST-108-078TS IST-108-115LR IST-108-115SR
731-0590EA 446 DKK
731-0590 731-0583 731-0587 731-0584 731-0588 731-0589 731-0591 731-0585 731-0586
Heat sealing foil, ThermASeal Foil™
Forsegling Mikrotiterpladetætninger
Peelable, strong bonding foil for polypropylene plates. Suitable for PCR, specifically water thermal cyclers, storage of solvents and other organics, including acids and alkaline, also for long term storage and transportation at low temperature.
  • Non pierceable
  • Ideal for water thermal cyclers
  • Good solvent resistance including DMSO
  • Seal integrity range: −200 to 110 °C