Hypodermic needles, Agani™
Leverandør: TERUMO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AN-2325R1 AN-2332R1 AN-2425R1 AN-2516R1 AN-2525R1 AN-2613R1 AN-2623R1 AN-2716R1 AN-2719R1 AN-3013R1 AN-1838R1 AN-1838S1 AN-1925R1 AN-1938R1 AN-1950R1 AN-2025R1 AN-2038R1 AN-2116R1 AN-2125R1 AN-2138R1 AN-2150R1 AN-2232R1 AN-2238R1 AN-2316R1
613-5382EA 61.3 DKK
613-5382 613-5381 613-5380 613-5379 613-5378 613-5377 613-5376 613-5375 613-5374 613-5373 613-5397 613-5396 613-5395 613-5394 613-5392 613-5391 613-5390 613-5389 613-5388 613-5387 613-5386 613-5385 613-5384 613-5383
Hypodermic needles, Agani™
Sprøjtenåle
These needles with thin wall are colour coded according to ISO.
  • Transparent needle hub for quick observation of blood reflux
  • Sterilised with ethylene oxide
  • Latex-free
  • Disposable
Emballering: Individual sterile blister packs with colour coding.