Sample vials, with screw cap, silanised glass
Leverandør: EP SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SAA-SV2-2 SAA-SV4-2 SCA-SV2-2 EP608154-S SCA-SV4-2 SCA-SV2-CT 212-01A-S EP608158-S C39-20A/CT-S C39-20C/CT-S
EPSCSAA-SV2-2EA 2530 DKK
EPSCSAA-SV2-2 EPSCSAA-SV4-2 EPSCSCA-SV2-2 EPSCEP608154-S EPSCSCA-SV4-2 EPSCSCA-SV2-CT EPSC212-01A-S EPSCEP608158-S EPSCC39-20A/CT-S EPSCC39-20C/CT-S
Sample vials, with screw cap, silanised glass
Hætteglas
These sample vials are mainly used to preserve the integrity of certain materials or extracts stored in glass containers.
  • Processing of materials prone to adhering to glass
  • Trace organic analysis and extraction glassware
  • Non-aqueous material storage
Surface deactivation treatments such as silanization may play an important role in preserving the integrity of certain materials or extracts stored in glass containers.
Pre-processed with a silanized surface deactivation treatment, these products are offered in convenient, ready-to-use clean packs.
It also helps to avoid alkalinization of contents which may occur as carbonates leach from the glass with normal weathering.
Leveringsinformation: Includes certificate of conformance.