Fastpek Allergen ELISA Kits
Leverandør: HYSERVE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1500005 1500007-2
SCIL1500005EA 2620 DKK
SCIL1500005 SCIL1500007-2
Fastpek Allergen ELISA Kits
Kliniske diagnose systemer Clinical Reagents and Kits
These kits are designed to detect allergens by special extraction buffer and antibodies.
  • High sensitivity
  • Increased specificity
  • Assay time of about 120 minutes
Suitable for use in highly processed food.