Laboratory coats, Hopen®
Leverandør: Medicom
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40.401/05 40.401/04 40.401/03
HOPE40.401/05EA 643 DKK
HOPE40.401/05 HOPE40.401/04 HOPE40.401/03
Laboratory coats, Hopen®
Engangsbeklædning
These disposable coats made of PP are ideal for use in non hazardous areas.
  • Lightweight PP is highly breathable
  • With collar and elasticated cuffs
  • Two pockets (one chest pocket, one side pocket)
  • Available with press-stud fastening or with zip closure
Typical applications include: food handling, research, laboratory work, maintenance and cleaning.
Weight: 40 g/m²