Allergen lateral flow IIR
Leverandør: HYSERVE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1500001
1500002
1500004
SCIL1500001EA
2800
DKK
SCIL1500001
SCIL1500002
SCIL1500004
Allergen lateral flow IIR
Mikrobiologiske testsystemer
Antigen-Antistof Test
Ag-AK, Lateral Flow
To detect allergen in highly processed foods by special extraction solution and unique antibody. This lateral-flow test is a high-performance qualitative assay.
- Easy to use and fast assay
- Available for various samples
- 20 sticks/kit