Overalls, Hopen®
Leverandør: Medicom
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40.101/04 40.101/05 40.101/03 40.101/06 40.101/02
HOPE40.101/04EA 611 DKK
HOPE40.101/04 HOPE40.101/05 HOPE40.101/03 HOPE40.101/06 HOPE40.101/02
Overalls, Hopen®
Engangsbeklædning
These disposable overalls with collar are made of breathable PP.
  • Lightweight polypropylene is highly breathable
  • With collar and elasticated cuffs and ankles
  • With zip closure (without flap)
Ideal for use in the chemical and petrochemical industry, for food applications, cleaning and non hazardous industrial applications.
Weight: 40 g/m²