Syringe filters, Life Sciences applications, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-1320 722-2520 723-2520 723-2545 723-2580 723-9920 723-9945 725-2520 725-2545 726-2520 726-2545
516-0930EA 3720 DKK
516-0930 516-0934 516-0935 516-0936 516-0937 516-0938 516-0939 516-0943 516-0944 516-0945 516-0946
Syringe filters, Life Sciences applications, Nalgene®
Filtre Sprøjtefiltre
These sterile syringe filters are ideal for sterilising (0,2 µm) or clarifying biological solutions, such as cell culture and microbial media, reagents and additives, proteinaceous solutions and buffers.
  • Three-year shelf life, with lot number and expiration date printed on individual syringe filter packages for easy traceability
  • Colour coded, with membrane and pore size printed on the syringe filter
  • Female Luer lock/male Luer slip inlet/outlet connection
PES membrane; PP housing: For sterile filtration and clarification of cell culture media, buffers and additives. Higher flow rates, lower protein binding and lower extractables than cellulosic membranes. Naturally hydrophilic; contains no wetting agents. Certified sterile, non-cytotoxic and non-pyrogenic.
SFCA membrane; PP housing: For cell culture fluids and cellular solutions. Low protein binding, lower extractables than standard CA.
CA membrane; PP housing: For sterile filtration and clarification of buffers, aqueous solutions and reagents. Some protein binding and extractables.
Nylon membrane; PP housing: Useful for alcohols and weak solvents. Naturally hydrophilic, low extractables, some protein binding.
CA membrane and integral glass fibre pre-filter, PP housing: These syringe filters are designed for filtering high particulate or viscous suspensions or solutions. Filter and pre-filter in one disposable unit. Certified non-cytotoxic and non-pyrogenic.
Emballering: Sterile, individual blister packed.