Anti-p23 Mouse Monoclonal Antibody [clone: JJ6]

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

18219-100 18219-25
CAYM18219-100EA 10770 DKK
CAYM18219-100 CAYM18219-25
Anti-p23 Mouse Monoclonal Antibody [clone: JJ6]
Antistoffer
Type: Primary
Antigen: p23
Clonality: Monoclonal
Clone: JJ6
Conjugation: unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Guinea Pig, Human, Mouse, Rabbit, S. Cerevisiae
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR