Anti-ALPI Sheep Polyclonal Antibody
Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
200-6135S 200-6135-0100
ROCK200-6135SEA 1690 DKK
ROCK200-6135S ROCK200-6135-0100
Anti-ALPI Sheep Polyclonal Antibody
Antistoffer
ALKALINE PHOSPHATASE antibody has been assayed against 1.0 ug of Alkaline Phosphatase [Calf Intestine] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG [H&L] (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000; Other Dilution: User Optimized
Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Bovine