Anti-H2AFZ Rabbit Polyclonal Antibody
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
140574 140574-100
USBI140574EA 3500 DKK
USBI140574 USBI140574-100
Anti-H2AFZ Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: H2AFZ
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,