Anti-Ly-6A/E Rat Monoclonal Antibody [clone: D7]

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L7715-05H1-50 L7715-05H1-500
USBIL7715-05H1-50EA 2070 DKK
USBIL7715-05H1-50 USBIL7715-05H1-500
Anti-Ly-6A/E Rat Monoclonal Antibody [clone: D7]
Antistoffer
Type: Primary
Antigen: Ly-6A/E
Clonality: Monoclonal
Clone: D7
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse,
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR