Bottles, Narrow Neck, Square
Leverandør: RIXIUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1-1201-0100-32 1-1201-0250-32 1-1201-0500-32 1-1201-1000-45 1-1201-010032W
RIXI1-1201-0100-32EA 1320 DKK
RIXI1-1201-0100-32 215-4748 215-4749 215-4750 215-4747
Bottles, Narrow Neck, Square
Flasker Glasflasker
Square bottles, soda-lime glass, amber, with dust protection cap, without screw caps. Amber coloured to protect light-sensitive materials.
  • Good chemical and physical properties
  • Stable, non-slip
  • For use in applications without high thermal loads
  • Screw caps to be ordered seperately