Ion selective electrode, nitrate, IntelliCAL™
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ISENO318101 ISENO318103 ISENO3181AP
HACHISENO318101EA 10250 DKK
HACHISENO318101 HACHISENO318103 HACHISENO3181AP
Ion selective electrode, nitrate, IntelliCAL™
Elektroder ISE elektroder
IntelliCAL™ ISENO3181 is a digital combination nitrate ion selective electrode (ISE) with a non refillable, double junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
The probe is available with a 1 or 3 meter cable and is intended for laboratory use. Ideal for measuring nitrate concentrations in drinking water, wastewater, and general water quality applications.
Bestillingsoplysninger: Electrode is supplied with integrated temperature sensor, calibration certificate, and basic user manual.
The analysis package contains the IntelliCAL™ ISENO318101 electrode, pack of nitrate ISA powder pillows (100), 1,0 mg/L nitrogen-nitrate standard solution (500 ml), 10 mg/L nitrogen-nitrate standard solution (500 ml), and 100 mg/L nitrogen-nitrate standard solution (500 ml).