Bottles, wide neck, with thread, without screw cap
Leverandør: EP SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
012-0120BP 012-0250BP 012-0500BP
EPSC012-0120BPEA 4250 DKK
EPSC012-0120BP EPSC012-0250BP EPSC012-0500BP
Bottles, wide neck, with thread, without screw cap
Flasker Plastflasker
HDPE, translucent.
  • Without screw caps
  • Wide mouth for easy filling and sample retrieval
  • Recommended for general purpose sampling where cleaning and certification is not required
  • Economically bulk packed without caps
Wide-mouth packers are general-purpose containers, bulk and unassembled. Unprocessed, ready for own cleaning procedure and assembly with screw caps purchased separately.