Filter plates, 850-DS 8-channel, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7707-3100 7707-3000 7707-3200 7707-3300 7707-3400 7707-3500 7707-3600 7707-3700 7707-3800 7707-3900
WHAT7707-3100EA 5270 DKK
WHAT7707-3100 WHAT7707-3000 WHAT7707-3200 WHAT7707-3300 WHAT7707-3400 WHAT7707-3500 WHAT7707-3600 WHAT7707-3700 WHAT7707-3800 WHAT7707-3900
Filter plates, 850-DS 8-channel, Whatman™
Mikroplader Filterplader
Whatman™ 850-DS 8-channel filter plates are specifically designed for use with the Agilent™ 850-DS Dissolution Sampling Station.
  • Automated processing of up to 8 samples simultaneously
  • Saves time
  • Eliminate errors associated with manual sampling
Developed in conjunction with Agilent™, Whatman™ 850-DS 8-channel filter plates increase productivity by allowing reliable alignment of the liquid path and reducing the risk of jamming or leaks that may occur with other dissolution sample preparation systems.