VWR® Dehydrated Culture Media in Jars
Leverandør: BDH Prolabo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6001.00900 6001.01125
6001.00900EA 1310 DKK
6001.00900 6001.01125 6134.05000 6012.05000
VWR® Dehydrated Culture Media in Jars
Mikrobiologiske medier Klargjort medium
For the preliminary, non selective enrichment of bacteria, particularly pathogenic enterobacteriaceae, from foodstuffs.
  • Convenient, just add water
  • Delivered in jars