Chemical resistant gloves, TevuChem® 765
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
765.10 765.11 765.8 765.9
112-4394EA 28590 DKK
112-4394 112-4395 112-4396 112-4397
Chemical resistant gloves, TevuChem® 765
Handsker
These two-layer gloves made of nitrile/nitrile foam and a cotton lining provide a excellent chemical resistance against aliphatic hydrocarbons and mineral oils.
  • Superior grip when handling wet and oily parts
  • Excellent chemical resistance and good mechanical properties
  • Very flexible, ergonomic fit
  • Latex-free
Ideal for use in oil production, petrochemical industries, metal processing industries and print shops.
Certificeringer: EN 388:2016 (3111X), EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5 VIRUS; Cat. III; AQL < 0,65