Cut resistant gloves, Nevocut® 923
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
923.7 923.8 923.9 923.10
KCHL923.7EA 3070 DKK
KCHL923.7 KCHL923.8 KCHL923.9 KCHL923.10
Cut resistant gloves, Nevocut® 923
Handsker
These seamless gloves combine the very good sensitivity, flexibility and wearing comfort of the knitted lining with maximum cut resistance (Level 5).
  • Glove is fully reversible
  • Can be washed several times without sacrificing its excellent fit
  • Excellent sensitivity, excellent sense of touch
The combination of polyamide, fiberglass and high-strength PE fibers offers a high level of cut resistance without restricting the mobility of the glove. This means that its is excellently suited to working with very sharp objects that also require great sensitivity. Likewise, its fine knitted structure means that it is perfect for wearing underneath another glove. Ideal for use in automotive, paper, glass and printing industries.
Certificeringer: EN 388:2016 (2X4XC), Cat. II
Food contact approved