Industrial gloves, Gobi® 112
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
112.10 112.9
KCHL112.10EA 2230 DKK
KCHL112.10 KCHL112.9
Industrial gloves, Gobi® 112
Handsker
This high quality cotton jersey liner with special nitrile coating meets all the requirements of a fully-coated protective glove with a wide usage spectrum: In addition to mechanical protection it also offers protection against liquids, high touch sensitivity and high temperature resistance.
  • Wide cuff for quick donning and doffing
  • Excellent mechanical resistance, moisture-repellent
  • Good grip when handling wet parts
  • Skin-friendly, high-quality cotton liner
  • Generous length for additional forearm protection
Ideal for metalworking and mechanical engineering, packaging and transportation works, building and construction, when working with oily and wet parts and for the plastics industry.
Certificeringer: EN 388 (3111), Cat. II