Crimp/snap top vials, Target® Snap-It™
Leverandør: THERMO NATIONAL SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C4011-5 C4011-73 C4011-LV2W
NSCAC4011-5EA 0 DKK
NSCAC4011-5 NSCAC4011-73 NSCAC4011-LV2W
Crimp/snap top vials, Target® Snap-It™
Hætteglas
These vials are made of superior quality 33 expansion borosilicate clear glass.
  • Wide opening, easier to fill and provides larger target area
  • Fits autosamplers that accomadates vials with 12×32 mm
  • Can be used with snap caps or aluminium crimp seal closures