Base materials for microbiology media
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2363.2 5210.2 8952.2 2363.1 2363.3 2365.1 2365.2 2365.3 2366.1 2366.2 2366.3 A157.1 A157.2 AE41.1 8952.1 8952.3 8986.1 8986.2 8986.3 2363.5 8986.4
ROTH2363.2EA 0 DKK
ROTH2363.2 ROTH5210.2 ROTH8952.2 ROTH2363.1 ROTH2363.3 ROTH2365.1 ROTH2365.2 ROTH2365.3 ROTH2366.1 ROTH2366.2 ROTH2366.3 ROTHA157.1 ROTHA157.2 ROTHAE41.1 ROTH8952.1 ROTH8952.3 ROTH8986.1 ROTH8986.2 ROTH8986.3 ROTH2363.5 ROTH8986.4
Base materials for microbiology media
Mikrobiologiske medier Base Materialer til Mikrobiologi
Peptone from soya beans is gained from papainic digestion of soya bean flour. Peptone tryptic digest is produced by proteolytic digestion of casein with trypsin-rich pancreas extract.
  • Granulated
  • Used for culture media, for fermentation and for the production of vaccines
  • Additive to nutrient media in the production of toxins, vaccins, enzymes and further biological products
  • Low-cost peptone