Culture media, ready to use, in tubes
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
14130-K 14131-K 14138-K 14140-K 14127-K 14137-K
SART14130-KEA 2080 DKK
SART14130-K SART14131-K SART14138-K SART14140-K 391-3998 801-4137
Culture media, ready to use, in tubes
Mikrobiologiske medier Klargjort medium
The traditional culture media for microorganisms is agar media. The media are available in two formats. Agar media in tubes are for pouring agar plates. The content of one tube is sufficient for two 90 mm or three 60 mm petri dishes.
  • A cost-effective alternative for casting plates