Safety shoes, lace-up, uvex mac crossroad, 6211, Heckel
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6211336 6211338 6211337 6211339 6211345 6211340 6211346 6211341 6211343 6211347 6211348
114-5905EA 0 DKK
114-5905 114-5907 114-5906 114-5908 114-5914 114-5909 114-5915 114-5910 114-5912 114-5916 114-5917
Safety shoes, lace-up, uvex mac crossroad, 6211, Heckel
Sko og støvler
100% metal-free multipurpose safety shoes made from liquid resistant full grain leather and a MACSOLE® rubber sole (resistant to oils, hydrocarbons and chemicals, heat resistant up to 300 °C).
  • Mesh lining
  • Removable, anatomically shaped CELL TECH hygienic insole: Perforated and shock absorbing
  • Two component rubber sole with excellent abrasion and cut resistance and self-cleaning profile
  • Rubber scuffcap, reinforced heel and sides
  • Metal-free, lightweight toecap (200 J)
  • Metal-free puncture resistant midsole
Suitable for uneven terrain and humid environments.
Colour: Black
Certificeringer: EN ISO 20345:2011; S3, CI, HI, HRO, SRC