Culture tubes, disposable, silanised
Leverandør: EP SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CTS-1275 CTS-16125-2 STT-13100-S STT-16100-S STT-16125-S CTS-13100 CTS-16100
EPSCCTS-1275EA 4480 DKK
EPSCCTS-1275 EPSCCTS-16125-2 EPSCSTT-13100-S EPSCSTT-16100-S EPSCSTT-16125-S EPSCCTS-13100 EPSCCTS-16100
Culture tubes, disposable, silanised
Rør Dyrkningsrør
Type 1 borosilicate glass, clear, with or without screw thread
  • Saves time and minimise the waste costs when performing quantitative analysis or storing materials
  • Avoid alkalinisation of contents which may occur as carbonates leach from the glass with normal weathering
These tubes are convenient and ready-to-use clean packs which are pre-processed with a silanised surface deactivation treatment which plays an important role in preserving the integrity of certain materials or extracts stored in glass containers.