Capillary pipettes, disposable, ringcaps®
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A764.1 A767.1
ROTHA764.1EA 999.82 DKK
ROTHA764.1 ROTHA767.1
Capillary pipettes, disposable, ringcaps®
Pipetter Mikrokapillærer
Borosilicate glass 3.3. These disposable capillary pipettes are autoclavable and come with a certificate of conformity.
  • Not heparinised
  • With ring mark
  • Tolerance to 5 µl 0,3%, 0,25%
  • Variation coefficient to 5 µl 0,60 %, from 10 µl 0,50%
Certificeringer: DIN ISO 7550; CE marked according to IVD 98/79 EG.