Protect (i)Xtreme Safety Ankle Boots, Lace-Up
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6246204/38 6246204/39 6246204/40 6246204/41 6246204/42 6246204/43 6246204/44 6246204/45 6246204/46 6246204/47 6246204/48
114-5903EA 1390 DKK
114-5903 114-5894 114-5895 114-5893 114-5892 114-5891 114-5890 114-5889 114-5888 114-5883 114-5881
Protect (i)Xtreme Safety Ankle Boots, Lace-Up
Sko og støvler
Non metallic full grain, leather ankle boot with a strong water resistant upper construction and an innovative PU/nitrile outsole for extrem performances.
  • (i)NTELLIGENT WEAR CONTROL: Helps users to detect when the outsole is used
  • Bi-density insole for improved comfort
  • Non metallic anti-puncture midsole
  • Smartium non metallic toecap made from injected polymer
Colour: Black.
Certificeringer: EN ISO 20345:2011; S3, SRC, HI, HRO, CI.