Culture media solutions, TransFast® system
Leverandør: DOEHLER GROUP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
204727782 204730001 204731782 440000000
710-0748EA 650 DKK
710-0748 710-0750 710-0853 710-1592
Culture media solutions, TransFast® system
Mikrobiologiske medier Klargjort medium
Fast and easy detection of beverage spoiling microorganisms. Qualitative detection of yeast, moulds, and lactic and acetic acid bacteria in beverages at pH <4,5 in under 48 hours.
  • Immediate application – ready-to-use broth and gel with no need to melt
  • Fast results – detection in under 48 hours instead of more than 96 hours
  • Broad sample spectrum – beverages, rinsing water, beverage raw materials, e.g. juice concentrates and membrane filtered samples
  • Simple screening – direct incubation and effortless monitoring using the TransFast® incubation lightbox
  • Increased efficiency – simple handling, early results, quick release