Chemical resistant gloves, Camatril® 733
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
733.8_U 733.9_U
KCHL733.8_UEA 0 DKK
KCHL733.8_U KCHL733.9_U
Chemical resistant gloves, Camatril® 733
Handsker
These unlined nitrile gloves provide high mechanical and chemical resistance. The high abrasion, cut and puncture resistance allows working with sharp edged materials.
  • Good grip when handling wet parts
  • Good temperature flexibility
  • Glove surface free from silicone (lacquer indifference, test method automotive industry)
  • Anti-static and electrostatic properties
  • Lower arm is protected by the long cuff
Ideal for use in agriculture, laboratories, in the automotive/petrochemistry/printing/chemical industry and in paint shops. Also suitable for the handling of epoxy resins.
Certificeringer: EN ISO 374-1 Type A (AJKLMO), EN 388:2016 (3102X); Cat. III; AQL <0,65