ESD safety shoes, slip-on, HACCP, Uni6, 31740
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31740/43 31740/42 31740/44 31740/35 31740/36 31740/46 31740/47 31740/48 31740/45 31740/38 31740/39 31740/40 31740/41 31740/37
114-5952EA 651 DKK
114-5952 114-5951 114-5953 114-5944 114-5945 114-5955 114-5956 114-5957 114-5954 114-5947 114-5948 114-5949 114-5950 114-5946
ESD safety shoes, slip-on, HACCP, Uni6, 31740
Sko og støvler
ESD safety shoe made from microfibre upper, with non slip TPU walking sole, which is resistant to animal fats, oils and fuels.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable acc Wave insole, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Shock-absorbent foam PU midsole
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for hygienically demanding areas.
Colour: White
Certificeringer: EN 61340 (ESD); DGUV 112-191; HACCP; EN ISO 20345:2011; S2, SRA