Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody
Leverandør: Genscript
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A01732-100 A01732-200
GENSA01732-100EA 4110 DKK
GENSA01732-100 GENSA01732-200
Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody
Antistoffer
Type: Tag
Antigen: NWSHPQFEK Tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu