Anti-NBS1 Mouse Monoclonal Antibody [clone: 7E4A2]
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NBP2-26297 NBP2-26297SS
NOVUNBP2-26297EA 3900 DKK
NOVUNBP2-26297 NOVUNBP2-26297SS
Anti-NBS1 Mouse Monoclonal Antibody [clone: 7E4A2]
Antistoffer
Type: Primary
Antigen: NBS1
Clonality: monoclonal
Clone: 7E4A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 Alpha
Reactivity: Human