Anti-ChABC Mouse Monoclonal Antibody [clone: 6A12]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NBP1-96142 NBP1-96142SS
NOVUNBP1-96142EA 4140 DKK
NOVUNBP1-96142 NOVUNBP1-96142SS
Anti-ChABC Mouse Monoclonal Antibody [clone: 6A12]
Antistoffer
Type: Primary
Antigen: ChABC
Clonality: monoclonal
Clone: 6A12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Pan
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR