Anti-BMI1 Mouse Monoclonal Antibody [clone: LLBmi1-1]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NBP1-96140 NBP1-96140SS
NOVUNBP1-96140EA 4240 DKK
NOVUNBP1-96140 NOVUNBP1-96140SS
Anti-BMI1 Mouse Monoclonal Antibody [clone: LLBmi1-1]
Antistoffer
Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: monoclonal
Clone: LLBmi1-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR