Anti-PIEZO2 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NBP1-78538 NBP1-78538SS
NOVUNBP1-78538EA 4600 DKK
NOVUNBP1-78538 NOVUNBP1-78538SS
Anti-PIEZO2 Rabbit Polyclonal Antibody
Antistoffer
The PIEZO2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIEZO2. This antibody reacts with human, mouse, porcine. The PIEZO2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: PIEZO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Porcine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR