Anti-NOTCH1 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NBP1-78292 NBP1-78292SS
NOVUNBP1-78292EA 4140 DKK
NOVUNBP1-78292 NOVUNBP1-78292SS
Anti-NOTCH1 Rabbit Polyclonal Antibody
Antistoffer
The Notch-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Notch-1. This antibody reacts with human, mouse. The Notch-1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: Notch-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR