Anti-BIRC3 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NBP1-27972 NBP1-27972SS
NOVUNBP1-27972EA 4140 DKK
NOVUNBP1-27972 NOVUNBP1-27972SS
Anti-BIRC3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BIRC3 (baculoviral IAP repeat containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR