Anti-NBS1 Rabbit Polyclonal Antibody
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NBP1-06705 NBP1-06705SS
NOVUNBP1-06705EA 4240 DKK
NOVUNBP1-06705 NOVUNBP1-06705SS
Anti-NBS1 Rabbit Polyclonal Antibody
Antistoffer
Rabbit Polyclonal NBS1 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: NBN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human