Anti-AGR2 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NBP1-05936 NBP1-05936SS
NOVUNBP1-05936EA 4140 DKK
NOVUNBP1-05936 NOVUNBP1-05936SS
Anti-AGR2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AGR2 (anterior gradient 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR