Anti-Ethenocytidine Mouse monoclonal antibody unconjugated [clone: 6F5]
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NB600-444 NB600-444SS
NOVUNB600-444EA 4140 DKK
NOVUNB600-444 NOVUNB600-444SS
Anti-Ethenocytidine Mouse monoclonal antibody unconjugated [clone: 6F5]
Antistoffer
Type: Primary
Antigen: Ethenocytidine
Clonality: monoclonal
Clone: 6F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 Lambda
Reactivity: Pan