Anti-HIST3H3 Rabbit Polyclonal Antibody
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NB21-1053 NB21-1053SS
NOVUNB21-1053EA 4570 DKK
NOVUNB21-1053 NOVUNB21-1053SS
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu