Anti-HIF1A Mouse Monoclonal Antibody [clone: HA111]
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NB100-296 NB100-296SS
NOVUNB100-296EA 4200 DKK
NOVUNB100-296 NOVUNB100-296SS
Anti-HIF1A Mouse Monoclonal Antibody [clone: HA111]
Antistoffer
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: HA111
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 Alpha
Reactivity: Human