Anti-CNX Rabbit Polyclonal Antibody
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NB100-1965 NB100-1965SS
NOVUNB100-1965EA 4360 DKK
NOVUNB100-1965 NOVUNB100-1965SS
Anti-CNX Rabbit Polyclonal Antibody
Antistoffer
The Calnexin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calnexin. This antibody reacts with human, mouse, rat, porcine, avian, bovine, chicken, drosophila, guinea pig, rabbit, sheep, xenopus, zebrafish. The Calnexin Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Simple Western,
Type: Primary
Antigen: CNX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Avian, Bovine, Chicken, Drosophila, Guinea Pig, Rabbit, Sheep, Xenopus, Zebrafish