Anti-EPAS1 Mouse monoclonal antibody unconjugated [clone: ep190b]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NB100-132 NB100-132SS
NOVUNB100-132EA 5490 DKK
NOVUNB100-132 NOVUNB100-132SS
Anti-EPAS1 Mouse monoclonal antibody unconjugated [clone: ep190b]
Antistoffer
Type: Primary
Antigen: EPAS1 (endothelial PAS domain protein 1)
Clonality: monoclonal
Clone: ep190b
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR