Anti-NTHL1 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NB100-108 NB100-108SS
NOVUNB100-108EA 4140 DKK
NOVUNB100-108 NOVUNB100-108SS
Anti-NTHL1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NTHL1 (nth endonuclease III-like 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR