Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NB100-105 NB100-105SS
NOVUNB100-105EA 5510 DKK
NOVUNB100-105 NOVUNB100-105SS
Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67]
Antistoffer
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: H1alpha67
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR