Anti-IgG Donkey Polyclonal Antibody (CF™ 543)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20314 20314-1
20314.EA 2680 DKK
20314. 20314-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 543)
Antistoffer
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR