Anti-IgG Goat Polyclonal Antibody (CF™ 647)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20280 20280-1
20280.EA 2680 DKK
20280. 20280-1
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 647 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR