Anti-IgG Donkey Polyclonal Antibody (CF™ 555)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20276 20276-1
20276.EA 2680 DKK
20276. 20276-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 555)
Antistoffer
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Guinea pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR