Anti-IgG2b Goat Polyclonal Antibody (CF™ 633)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20270 20270-1
20270.EA 2870 DKK
20270. 20270-1
Anti-IgG2b Goat Polyclonal Antibody (CF™ 633)
Antistoffer
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse